Rubbermaid® Commercial Bouncer® Measuring Cups, Essendant
Supplier: Janitorial Supplies
RCP3215CLE RCP3217CLE RCP3216CLE RCP3218CLE
89073-196EA 54.46 USD
89073-196 89073-200 89073-198 89073-202
Rubbermaid® Commercial Bouncer® Measuring Cups, Essendant
Beakers
Rubbermaid measuring cups feature large, easy-to-read measurements in high-contrast red or blue.
  • Constructed from break-resistant polycarbonate
  • Easy-to-read measurements in high-contrast blue or red.
  • For use with temperatures ranging from –40 to +1 °F
  • Measures liquid or dry ingredients to help save food costs
  • Made in the USA
Measuring cup is constructed of break-resistant and durable crystal-clear material and commercial dishwasher safe.
Certificaciones: USDA approved, certified to NSF standard #2.