Acrylamide ≥99.9%, crystalline powder, Ultrapure for electrophoresis

Supplier: MP BIOMEDICALS (FKA ICN BIOMED
Danger

Synonyms: Prop-2-enamide

04814340
IC814340EA 113.8 USD
IC814340
Acrylamide ≥99.9%, crystalline powder, Ultrapure for electrophoresis
Acrylamide (monomer)

Acrylic Acid Content <0.01%


Acrylamide is a monomer used in a variety of synthetic processes to form polymers and copolymers. It polymerizes in the presence of free radicals in aqueous solutions. It is usually used to prepare polyacrylamide gels for electrophoresis separation of biomolecules. Common reaction initiators are riboflavin and ammonium persulfate; varying the ratio of acrylamide to crosslinking agent permits the formation of a gel with predictable average pore size and texture. Acrylamide tends to hydrolyze under acidic or basic conditions to form acrylic acid. Polyacrylamide plastics have numerous commercial applications, due in part to the tendency of polyacrylamide to adsorb many times its own mass in water. Acrylic acid and any ionic impurities in the acrylamide can have significant effects on the performance of the acrylamide gel formed, since the voltage across the gel is affected by the ionic charge of the gel and usage buffers.


Soluble in water (1% aq soln-Clear, Colorless Solution), methanol (155 g/100 mL), ethanol (86 g/100 mL), alcohol, acetone (63.1 g/100 mL); only minimally soluble in benzene or heptane.

Formula: C₃H₅NO
MW: 71.08 g/mol
Boiling Pt: 232 °C (1013 hPa)
Melting Pt: 82…92 °C
Density: 1.122 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Ambient
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://pr.vwr.com/stibo/web/std.lang.all/13/21/2301321.jpg

Order Now

Specification Test Results

Identity Test Passes
Purity >99.9%
FTIR Conforms to Standard
Melting Point 82 - 92 °C
OD290 (1% aq soln) <0.10
Acrylic Acid Content <0.001%
pH 5.0 - 7.0 (1% aq soln)
Solubility (40% aq soln) Clear, Colorless Solution

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR