n-Nonyl-β-D-maltoside ≥99.5%, Crystallization grade
Supplier: CUBE BIOTECH
Synonyms:
N-Tridecyl Beta-Maltoside, NM
16065
16066
16067
77392-398PK
405.82
USD
77392-398
77392-400
77392-402
n-Nonyl-β-D-maltoside ≥99.5%, Crystallization grade
n-Nonyl-β-D-maltoside
Mild, non-ionic detergents such as Nonylmaltoside are well suited for the solubilization, stabilization, and purification of membrane proteins. For screening of the best-suited detergent for a given application, we offer maltosides with chain lengths from C-9 to C-13.
Molecular Weight: 468.41 g/mol
Form: White crystalline powder
CMC (in H2O): 6 mM
Formula:
C₂₁H₄₀O₁₁ MW: 468.54 g/mol |
MDL Number:
MFCD08062400 CAS Number: 106402-05-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...