HighSpec Rho1D4 MagBeads XL
Supplier: CUBE BIOTECH
HighSpec Rho1D4 MagBeads XL is a suspension of 90 µm magnetic agarose beads coupled to an anti-Rho1D4 antibody.
- Usage: Specific binding and purification of Rho1D4-tagged proteins
- Specificity: Affinity to Rho1D4-tagged (TETSQVAPA) proteins
- Binding capacity: >3 mg/ml
- Bead Ligand: anti-Rho1D4 antibody
- Bead size: 90 μm
- Matrix: 6% cross-linked agarose
- Filling quantity: Delivered as a 25% suspension
- Short-term storage: In neutral buffer at 4 °C
- Long-term storage: In neutral buffer with 20% ethanol at 4 °C
HighSpec Rho1D4 MagBeads XL resin is meant for protein purification assays of the highest possible purity, exploiting the specificity of an antibody to its antigen. Additionally, we offer Rho1D4-tag affinity resin with a bead diameter of 30 µm for more mechanical stability and slightly higher protein yield. Alternatively, Rho1D4 Agarose resin is also available if you prefer this affinity chromatography method. Additional support products for Rho1D4-tag purification assays are Rho1D4 peptide, an essential compound of the elution buffer, and our Rho1D4 antibody for later western blot detection of your protein.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...