Cylinder and Bottle Brushes, Radial End, Mill-Rose
Supplier: Mill-Rose
72064B 72065B 3065B 72063B
17066-058PK 45.06 USD
17066-058 17066-080 17066-089 70230-018
Cylinder and Bottle Brushes, Radial End, Mill-Rose
Brushes
For washing cylinders, large test tubes and bottles.
  • Twisted wire handle with hanging loop
  • Full radial end provides thorough cleaning action
Available in black natural bristles or black nylon filaments.