PureCube NHS Activated MagBeads

Supplier: CUBE BIOTECH

50405 50425 50490 50491 50401
77393-184EA 804.43 USD
77393-184 77393-186 77393-188 77393-190 77393-182
PureCube NHS Activated MagBeads
Protein Purification Protein Purification Magnetic Beads

PureCube NHS (N-hydroxy succinimide) MagBeads/magnetic beads are your best option to couple biomolecules with free-standing amine groups.


  • Usage: Coupling proteins to active NHS Groups
  • Specificity: Exposed amine (NH₂) groups on biomolecule
  • Binding capacity: >25 mg/ml
  • Bead Ligand: Chemically active NHS groups
  • Bead size: 25 to 30 μm
  • Filling quantity: Delivered as a 25% suspension
  • Storage Buffer: 100% isopropanol
  • Matrix: 6% cross-linked agarose
  • Storage Temperature: 4 °C
  • Shelf life: If properly stored - 6 months upon delivery, unopened
  • Chemical Structure: NHS structure
  • Special Notes: This product is shipped as a flammable good, 2-propanol UN1219


The resulting Amide bond between the NHS MagBeads and your biomolecule of interest is stable so that the bead can be used for other applications afterward. Our PureCube NHS Activated MagBeads/magnetic beads are also available as agarose resin for FPLC or as XL-sized MagBeads.

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR