QuadMag Genie®, 230 V
Supplier: Electron Microscopy Sciences
72402-20 72402-AU 72402-BR 72402-EU 72402-SW
102098-018EA 1901.71 USD
102098-018 102098-020 102098-022 102098-024 102098-026
QuadMag Genie®, 230 V
Stirrers Magnetic Stirrers, Multi-position
QuadMag Genie® is a general purpose multi-position magnetic stirrer designed for general laboratory applications which provides vigorous stirring for up to 4 vessels simultaneously.
  • 230 V model
The powerful magnet and motor offer exceptional magnetic force capable of stirring up to 2 L for each position. Powerful 'rare earth' magnets automatically pull stir bars into position and stay coupled even under the most demanding stirring conditions. Rugged ABS/Polycarbonate housing and the famous Vortex-Genie® heavy-duty motor provide years of reliable service. Spill-resistant housing channels fluids away from the internal components.
Precaución: 230 V model. Corresponding model with voltage suitable for use in US (102098-016) is also available, please see separate product page.