MultiMagStir Genie Magnetic Stirrers, 230 V
Supplier: Electron Microscopy Sciences
72405-20 72405-AU 72405-BR 72405-EU
102098-040EA 1757.56 USD
102098-040 102098-042 102098-044 102098-046
MultiMagStir Genie Magnetic Stirrers, 230 V
Stirrers Magnetic Stirrers
  • 230 V model
  • The revolutionary four position MultiMagStir Genie provides low to high speed stirring and editable memory profiles
  • Microprocessor controls offer precision and reproducibility without internal heat build-up
  • Customize your application with the four-position MultiMagStir Genie!
  • MultiMagStir Genie may be used with up to four 1 L flasks/beakers or three 1 L spinner flasks and a maximum sample weight of 10.0 kg
Speed range of 10 to 1200 rpm including reverse and interval stirring for applications ranging from gentle stirring for cell culture to aggressive mixing for viscous polymers. Three power levels are available for various sample viscosities. Low-profile magnetic stirrer uses microprocessor control for precise and reproducible operation without heat build-up from internal friction. Manual mode provides stir, stir reverse, and time functions. Profiles mode allows users to quickly program speed, ramp rate, stir direction, time (including pause), and power level in up to nine distinct steps. Up to four stirring programs can be saved. Automatic mode recalls and executes one of up to four user-defined protocols.
Precaución: 230 V model. Corresponding model with voltage suitable for use in US (102098-038) is also available, please see separate product page.