Corning® digital stirring Hot Plates use innovative microprocessor technology with digital display which optimally controls the heating and stirring to the Pyroceram glass-ceramic stirring hot plates.
- 220 V model
- Closed-loop heating and stirring controls that monitor and regulate both temperature and stirring speed
- Microprocessor maintains consistent and repeatable temperature setting from 5 °C (if ambient temperature is 0 °C or lower) up to 550 °C
- Digital LED temperature display is adjustable in 5 °C increments and blinks until temperature is reached
- Optional external temperature controller truly controls temperature inside the vessel and eliminates the need to closely monitor liquid temperature
- Exclusive closed-loop stirring control monitors and regulates the stirring speed from 60 to 1150 RPM for aqueous, viscous, or semi-solids solutions
- Digital LED speed display gives you better control of your stirring process
- Stronger, larger magnet improves stir bar coupling combined with automatic up speed adjustment minimizes decoupling
- Stronger, larger magnet improves stir bar coupling combined with automatic up speed adjustment minimizes decoupling
For safety there is a bright LED HOT TOP icon that lights up when top plate is over 60 °C, even when heat control is turned off. Separate temperature sensor provides power cutoff if unit overheats and an optional temperature controller will shut off unit when is not in liquid. A two piece casting design deflects spills away from electronic and includes a built in mount for an optional 18'' (45.7 cm) support rod.
Certificaciones: UL and cUL approved.
Precaución: 220 V model. Corresponding model with voltage suitable for use in US (100493-718) is also available, please see separate product page.