Tris(3-fluoropropyl)borate

Supplier: APOLLO SCIENTIFIC
Warning

PC450192-1G PC450192-250MG PC450192-5G
77415-480EA 405.48 USD
77415-480 77415-482 77415-484
Tris(3-fluoropropyl)borate
Tris(3-fluoropropyl)borate
Formula: C₉H₁₈BF₃O₃
Storage Temperature: Ambient
MDL Number: MFCD20489408
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR