3-(Pentafluorothio)benzylamine

Supplier: APOLLO SCIENTIFIC
Danger

PC303050-1G PC303050-5G
77414-350EA 122.2 USD
77414-350 77414-352
3-(Pentafluorothio)benzylamine
3-(Pentafluorothio)benzylamine
Formula: C₇H₈F₅NS
Storage Temperature: Ambient
MDL Number: MFCD06213674
CAS Number: 771573-34-3
UN: 2735
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR