2-Chloro-5-(difluoromethoxy)benzyl bromide 95%

Supplier: APOLLO SCIENTIFIC
Danger

PC302227-10G PC302227-1G PC302227-5G
77414-138EA 28.32 USD
77414-138 77414-140 77414-142
2-Chloro-5-(difluoromethoxy)benzyl bromide 95%
2-Chloro-5-(difluoromethoxy)benzyl bromide
Formula: C₈H₆BrClF₂O
Storage Temperature: Ambient
MDL Number: MFCD09832368
CAS Number: 1017779-54-2
UN: 3265
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR