4-Hydroxy-7-methylthieno[3,2-d]pyrimidine

Supplier: APOLLO SCIENTIFIC
Warning

OR110039-1G OR110039-5G
77403-030EA 28.56 USD
77403-030 77403-032
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine
4-Hydroxy-7-methylthieno[3,2-d]pyrimidine
Formula: C₇H₆N₂OS
Melting Pt: 240…242 °C
Storage Temperature: Ambient
MDL Number: MFCD13181517
CAS Number: 175137-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR