Guanosine

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-Amino-9-((2R,3R,4S,5R)-3,4-dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-3H-purin-6(9H)-one

OR4880-100G OR4880-25G OR4880-500G
77410-064EA 52.98 USD
77410-064 77410-066 77410-068
Guanosine
Guanosine
Formula: C₁₀H₁₃N₅O₅
MW: 283.24 g/mol
Melting Pt: 250 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00010182
CAS Number: 118-00-3
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR