6-Hydroxypicolinic acid 95%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 6-Hydroxy-2-pyridinecarboxylic acid, 6-Hydroxypyridine-2-carboxylic acid

OR1911-100G OR1911-25G
77406-434EA 182.36 USD
77406-434 77406-436
6-Hydroxypicolinic acid 95%
6-Hydroxypicolinic acid
Formula: C₆H₅NO₃
MW: 139.11 g/mol
Melting Pt: 270 °C
Storage Temperature: Ambient
MDL Number: MFCD00192220
CAS Number: 19621-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR