Tin(II) chloride, anhydrous 98%

Supplier: APOLLO SCIENTIFIC
Danger

IN3644-100G IN3644-500G
77401-576EA 77 USD
77401-576 77401-578
Tin(II) chloride, anhydrous 98%
Tin(II) chloride
Formula: SnCl₂
Boiling Pt: 652 °C
Melting Pt: 247 °C
Density: 3.95 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011241
CAS Number: 7772-99-8
EINECS: 231-868-0
UN: 3260
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR