6-Aminopyridin-2-ol, Technical Grade

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 6-Hydroxypyridin-2-amine, 6-Aminopyridin-2-ol

OR4269-1G OR4269-25G OR4269-5G
77409-674EA 21.32 USD
77409-674 77409-676 77409-678
6-Aminopyridin-2-ol, Technical Grade
6-Aminopyridin-2-ol
Formula: C₅H₆N₂O
MW: 110.11 g/mol
Melting Pt: 210…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00039717
CAS Number: 59315-47-8
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR