2-Fluoro-3-methoxybenzoyl chloride 97%
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
3-(Chlorocarbonyl)-2-fluoroanisole
PC9296-5G
PC9296-1G
PC9296-25G
77418-654EA
158.94
USD
77418-654
77418-650
77418-652
2-Fluoro-3-methoxybenzoyl chloride 97%
2-Fluoro-3-methoxybenzoyl chloride
Formula:
C₈H₆ClFO₂ MW: 188.583 g/mol Boiling Pt: 133 °C (12 mm Hg) Melting Pt: 49…51 °C Storage Temperature: Ambient |
MDL Number:
MFCD08277634 CAS Number: 850563-45-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...