2,3-Difluoro-6-nitrotoluene 95%

Supplier: APOLLO SCIENTIFIC

PC902026-10G PC902026-1G PC902026-5G
77418-504EA 284.24 USD
77418-504 77418-506 77418-508
2,3-Difluoro-6-nitrotoluene 95%
2,3-Difluoro-6-nitrotoluene
Formula: C₇H₅F₂NO₂
Storage Temperature: Ambient
MDL Number: MFCD08706407
CAS Number: 914348-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR