[2-(Perfluorodec-1-yl)ethyl]trichlorosilane 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Henicosafluorododecyltrichlorosilane, 1-[2-(Trichlorosilyl)ethyl]perfluorodecane

PC8947-1G PC8947-5G
77418-488EA 69.28 USD
77418-488 77418-490
[2-(Perfluorodec-1-yl)ethyl]trichlorosilane 97%
[2-(Perfluorodec-1-yl)ethyl]trichlorosilane
Formula: C₁₂H₄Cl₃F₂₁Si
MW: 681.571 g/mol
Boiling Pt: 113…114 °C (4 mm Hg)
Melting Pt: 50…55 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07368770
CAS Number: 102488-49-3
UN: 2987
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR