4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pent-2-enoic acid 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-(Perfluoroisopropyl)acrylic acid, 3-(Heptafluoroprop-2-yl)prop-2-enoic acid

PC6932L-1G PC6932L-25G PC6932L-5G
77417-604EA 34.64 USD
77417-604 77417-606 77417-608
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pent-2-enoic acid 95%
4,5,5,5-Tetrafluoro-4-(trifluoromethyl)pent-2-enoic acid
Formula: C₆H₃F₇O₂
MW: 240.076 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09998187
CAS Number: 243139-64-2
UN: 3261
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR