Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic acid 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: Perfluoro-2,5-dimethyl-3,6-dioxa-8-propoxynonanoic acid

PC6851-1G PC6851-5G
77417-558EA 48.9 USD
77417-558 77417-560
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic acid 95%
Perfluoro-2,5,8-trimethyl-3,6,9-trioxadodecanoic acid
Formula: C₁₂HF₂₃O₅
MW: 662.097 g/mol
Boiling Pt: 75…77 °C
Density: 1.8 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00236739
CAS Number: 65294-16-8
UN: 3265
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR