2-Chloro-3-fluoro-6-nitroaniline 97%

Supplier: APOLLO SCIENTIFIC
Danger

PC6781-1G PC6781-5G
77417-530EA 27.2 USD
77417-530 77417-532
2-Chloro-3-fluoro-6-nitroaniline 97%
2-Chloro-3-fluoro-6-nitroaniline
Formula: C₆H₄ClFN₂O₂
Melting Pt: 127…129 °C
Storage Temperature: Ambient
MDL Number: MFCD09763600
CAS Number: 948014-34-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR