1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carbonyl chloride 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2-[4-(Chlorocarbonyl)piperidin-1-yl]-4-(trifluoromethyl)pyrimidine,4-(Chlorocarbonyl)-1-[4-(trifluoromethyl)pyrimidin-2-yl]piperidine

PC6733-1G PC6733-250MG
77417-520EA 192.56 USD
77417-520 77417-522
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carbonyl chloride 97%
1-[4-(Trifluoromethyl)pyrimidin-2-yl]piperidine-4-carbonyl chloride
Formula: C₁₁H₁₁ClF₃N₃O
MW: 293.673 g/mol
Boiling Pt: 158 °C (2 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD02091616
CAS Number: 306934-79-2
UN: 1759
ADR: 8,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR