Tris(perfluorohex-1-yl)amine, tech
Supplier: APOLLO SCIENTIFIC
|
Danger
|
Synonyms:
Tris(tridecafluorohex-1-yl)amine
PC6220G-1G
PC6220G-5G
77417-314EA
75.4
USD
77417-314
77417-316
Tris(perfluorohex-1-yl)amine, tech
Tris(perfluorohex-1-yl)amine
Formula:
C₁₈F₃₉N MW: 971.137 g/mol Boiling Pt: 250…260 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00042590 CAS Number: 432-08-6 UN: 3261 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...