5-Amino-2-(methylsulfonyl)benzotrifluoride

Supplier: APOLLO SCIENTIFIC
Warning

PC6125-1G PC6125-250MG PC6125-5G
77417-288EA 49.28 USD
77417-288 77417-290 77417-292
5-Amino-2-(methylsulfonyl)benzotrifluoride
5-Amino-2-(methylsulfonyl)benzotrifluoride
Formula: C₈H₈F₃NO₂S
Melting Pt: 141…141 °C
Storage Temperature: Ambient
MDL Number: MFCD12807191
CAS Number: 252561-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR