3-Perfluorohexyl-2-iodopropan-1-ol 97%

Supplier: APOLLO SCIENTIFIC
Warning

PC6088D-1G PC6088D-5G
77417-278EA 108.4 USD
77417-278 77417-280
3-Perfluorohexyl-2-iodopropan-1-ol 97%
3-Perfluorohexyl-2-iodopropan-1-ol
Formula: C₉H₆F₁₃IO
Boiling Pt: 90 °C (1.5 mm Hg)
Melting Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00236070
CAS Number: 38550-44-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR