1H,1H,2H,2H-Perfluorododecyl acrylate 97%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 2-(Perfluorodec-1-yl)ethyl acrylate, 3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12,12-Henicosafluorododecyl prop-2-enoate

PC5969O-100G PC5969O-25G PC5969O-5G
77417-222EA 646.94 USD
77417-222 77417-224 77417-226
1H,1H,2H,2H-Perfluorododecyl acrylate 97%
1H,1H,2H,2H-Perfluorododecyl acrylate
Formula: C₁₅H₇F₂₁O₂
Boiling Pt: 122 °C (4 mm Hg)
Melting Pt: 48…53 °C
Density: 1.32 g/cm³ (25 °C)
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00236108
CAS Number: 17741-60-5
EINECS: 241-732-2
UN: 3077
ADR: 9+EHS,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR