2-Fluoro-4-methylphenylacetonitrile 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: 2-(2-Fluoro-4-methylphenyl)acetonitrile

PC5937-25G PC5937-5G PC5937-1G
77417-194EA 583.78 USD
77417-194 77417-196 77417-192
2-Fluoro-4-methylphenylacetonitrile 97%
2-Fluoro-4-methylphenylacetonitrile
Formula: C₉H₈FN
Storage Temperature: Ambient
MDL Number: MFCD03094328
CAS Number: 518070-26-3
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR