4-Bromo-2-(trifluoromethyl)phenylacetylene 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC56843-1G PC56843-250MG
77416-914EA 96.78 USD
77416-914 77416-916
4-Bromo-2-(trifluoromethyl)phenylacetylene 95%
4-Bromo-2-(trifluoromethyl)phenylacetylene
Formula: C₉H₄BrF₃
Storage Temperature: Refrigerator
MDL Number: MFCD31654734
CAS Number: 2244085-78-5
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR