3-Bromo-2-fluorophenylacetylene 98%

Supplier: APOLLO SCIENTIFIC
Warning

PC56837-1G PC56837-250MG
77416-888EA 181.34 USD
77416-888 77416-890
3-Bromo-2-fluorophenylacetylene 98%
3-Bromo-2-fluorophenylacetylene
Formula: C₈H₄BrF
Storage Temperature: Refrigerator
MDL Number: MFCD31654731
CAS Number: 1701568-15-1
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR