Difluoromethanesulfonyl chloride

Supplier: APOLLO SCIENTIFIC
Danger

PC53298-1G PC53298-250MG PC53298-5G
77416-668EA 187.46 USD
77416-668 77416-670 77416-672
Difluoromethanesulfonyl chloride
Difluoromethanesulfonyl chloride
Formula: CHClF₂O₂S
MW: 150.532 g/mol
Boiling Pt: 95…99 °C
Density: 1.696 g/cm³ (25 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD09971548
CAS Number: 1512-30-7
EINECS: 216-150-7
UN: 2924
ADR: 3+8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR