2,2,3,3,4,4,4-Heptafluorobutyl chloroformate 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 2,2,3,3,4,4,4-Heptafluorobutyl carbonochloridate

PC53244-1G PC53244-5G
77416-658EA 131.42 USD
77416-658 77416-660
2,2,3,3,4,4,4-Heptafluorobutyl chloroformate 97%
2,2,3,3,4,4,4-Heptafluorobutyl chloroformate
Formula: C₅H₂ClF₇O₂
MW: 262.51 g/mol
Boiling Pt: 108…109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD25955259
CAS Number: 108156-21-4
UN: 3277
ADR: 6.1+8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR