2,3-Difluoro-5-ethoxytoluene 97%

Supplier: APOLLO SCIENTIFIC

Synonyms: 5-Ethoxy-1,2-difluoro-3-methylbenzene

PC52479-1G PC52479-5G
77416-556EA 124.3 USD
77416-556 77416-558
2,3-Difluoro-5-ethoxytoluene 97%
2,3-Difluoro-5-ethoxytoluene
Formula: C₉H₁₀F₂O
MW: 172.172 g/mol
Storage Temperature: Ambient
MDL Number: MFCD28788825
CAS Number: 1803790-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR