2,6-Dibromo-1-(pentafluorosulfur)benzene 95%

Supplier: APOLLO SCIENTIFIC

PC52458-1G PC52458-250MG PC52458-5G
77416-550EA 266.6 USD
77416-550 77416-552 77416-554
2,6-Dibromo-1-(pentafluorosulfur)benzene 95%
2,6-Dibromo-1-(pentafluorosulfur)benzene
Formula: C₆H₃Br₂F₅S
Storage Temperature: Ambient
MDL Number: MFCD32708477
CAS Number: 2514942-08-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR