4-Chloro-3-(trifluoromethyl)benzyl chloride 98%

Supplier: APOLLO SCIENTIFIC

PC520528-1G PC520528-25G PC520528-5G
77416-494EA 49.92 USD
77416-494 77416-496 77416-498
4-Chloro-3-(trifluoromethyl)benzyl chloride 98%
4-Chloro-3-(trifluoromethyl)benzyl chloride
Formula: C₈H₅Cl₂F₃
Storage Temperature: Ambient
MDL Number: MFCD03412205
CAS Number: 23131-73-9
UN: 1760
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR