N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride 92%

Supplier: APOLLO SCIENTIFIC
Danger

PC520402-100MG PC520402-1G PC520402-250MG
77416-484EA 21.26 USD
77416-484 77416-486 77416-488
N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride 92%
N-[1-(2-Fluorophenyl)ethyl]prop-2-yn-1-amine hydrochloride
Formula: C₁₁H₁₂FN·ClH
MW: 213.679 g/mol
Storage Temperature: Freezer
MDL Number: MFCD31745724
CAS Number: 2379918-37-1
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR