4-(2,2,2-Trifluoroethoxy)aniline 98%

Supplier: APOLLO SCIENTIFIC
Warning

Synonyms: 4-Amino-β,β,β-trifluorophenetole

PC5198-1G PC5198-5G
77416-474EA 24.46 USD
77416-474 77416-476
4-(2,2,2-Trifluoroethoxy)aniline 98%
4-(2,2,2-Trifluoroethoxy)aniline
Formula: C₈H₈F₃NO
MW: 191.15 g/mol
Melting Pt: 63…66 °C
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00077606
CAS Number: 57946-61-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR