3-Bromo-5-fluorophenylisocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503012-1G PC503012-5G
77416-316EA 75.4 USD
77416-316 77416-318
3-Bromo-5-fluorophenylisocyanate
3-Bromo-5-fluorophenylisocyanate

May form small amount of sediment.

Formula: C₇H₃BrFNO
Storage Temperature: Refrigerator
MDL Number: MFCD11619406
CAS Number: 1094485-14-9
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR