3-Bromo-4-(trifluoromethoxy)phenylisocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503008-25G PC503008-5G PC503008-1G
77416-294EA 63.16 USD
77416-294 77416-296 77416-292
3-Bromo-4-(trifluoromethoxy)phenylisocyanate
3-Bromo-4-(trifluoromethoxy)phenylisocyanate

May form small amount of sediment.

Formula: C₈H₃BrF₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32200431
CAS Number: 685859-17-0
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR