3-Bromo-4-(trifluoromethoxy)phenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503007-1G PC503007-25G PC503007-5G
77416-286EA 83.54 USD
77416-286 77416-288 77416-290
3-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
3-Bromo-4-(trifluoromethoxy)phenylisothiocyanate
Formula: C₈H₃BrF₃NOS
MW: 298.08 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32200430
CAS Number: 1379677-28-7
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR