3-Bromo-2-fluorophenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503005-1G PC503005-25G PC503005-5G
77416-274EA 79.46 USD
77416-274 77416-276 77416-278
3-Bromo-2-fluorophenylisothiocyanate
3-Bromo-2-fluorophenylisothiocyanate
Formula: C₇H₃BrFNS
MW: 232.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD26686203
CAS Number: 1517289-83-6
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR