2-Bromo-6-fluorophenylisothiocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC503002-1G PC503002-25G PC503002-5G
77416-256EA 83.54 USD
77416-256 77416-258 77416-260
2-Bromo-6-fluorophenylisothiocyanate
2-Bromo-6-fluorophenylisothiocyanate
Formula: C₇H₃BrFNS
MW: 232.073 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22557646
CAS Number: 1360888-03-4
UN: 2922
ADR: 8+6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR