2-Bromo-4-(trifluoromethoxy)phenylisocyanate

Supplier: APOLLO SCIENTIFIC
Danger

PC502997-1G PC502997-25G PC502997-5G
77416-226EA 21.26 USD
77416-226 77416-228 77416-230
2-Bromo-4-(trifluoromethoxy)phenylisocyanate
2-Bromo-4-(trifluoromethoxy)phenylisocyanate

May form small amount of sediment.

Formula: C₈H₃BrF₃NO₂
Storage Temperature: Refrigerator
MDL Number: MFCD32200433
CAS Number: 848771-80-2
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR