2,5-Difluoro-1-(trifluoromethoxy)benzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC501501-1G PC501501-250MG PC501501-5G
77416-074EA 65.28 USD
77416-074 77416-076 77416-078
2,5-Difluoro-1-(trifluoromethoxy)benzene 99%
2,5-Difluoro-1-(trifluoromethoxy)benzene
Formula: C₇H₃F₅O
Storage Temperature: Ambient
MDL Number: MFCD22580924
CAS Number: 1404195-06-7
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR