2,3-Difluoro-1-(trifluoromethoxy)benzene 99%

Supplier: APOLLO SCIENTIFIC
Warning

PC501494-1G PC501494-25G PC501494-5G
77416-062EA 30.56 USD
77416-062 77416-064 77416-066
2,3-Difluoro-1-(trifluoromethoxy)benzene 99%
2,3-Difluoro-1-(trifluoromethoxy)benzene
Formula: C₇H₃F₅O
MW: 198.09 g/mol
Storage Temperature: Ambient
MDL Number: MFCD18641903
CAS Number: 158178-35-9
UN: 1993
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR