2-Bromo-3-fluoro-6-nitrophenol 95%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 3-Bromo-4-fluoro-2-hydroxynitrobenzene, 2-Bromo-1-fluoro-3-hydroxy-4-nitrobenzene

PC49004-1G PC49004-25G PC49004-5G
77415-836EA 29.4 USD
77415-836 77415-838 77415-840
2-Bromo-3-fluoro-6-nitrophenol 95%
2-Bromo-3-fluoro-6-nitrophenol
Formula: C₆H₃BrFNO₃
Melting Pt: 65…66 °C
Storage Temperature: Ambient
MDL Number: MFCD18071060
CAS Number: 103979-08-4
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR