2-(Difluoromethyl)benzonitrile

Supplier: APOLLO SCIENTIFIC
Warning

PC48525-5G PC48525-1G
77415-774EA 160.98 USD
77415-774 77415-772
2-(Difluoromethyl)benzonitrile
2-(Difluoromethyl)benzonitrile
Formula: C₈H₅F₂N
Storage Temperature: Ambient
MDL Number: MFCD21606585
CAS Number: 799814-30-5
UN: 3276
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR