Ethyl 2,2-difluorohexanoate 95%

Supplier: APOLLO SCIENTIFIC
Warning

PC48304-1G PC48304-250MG PC48304-5G
77415-744EA 32.44 USD
77415-744 77415-746 77415-748
Ethyl 2,2-difluorohexanoate 95%
Ethyl 2,2-difluorohexanoate
Formula: C₈H₁₄F₂O₂
Boiling Pt: 83…85 °C (15 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD11007687
CAS Number: 74106-81-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR