2,3-Dihydro-8-fluoroquinolin-4(1H)-one 97%

Supplier: APOLLO SCIENTIFIC
Danger

Synonyms: 8-Fluoro-4-oxo-1,2,3,4-tetrahydroquinoline

PC4676-1G PC4676-250MG
77415-666EA 326.02 USD
77415-666 77415-668
2,3-Dihydro-8-fluoroquinolin-4(1H)-one 97%
2,3-Dihydro-8-fluoroquinolin-4(1H)-one
Formula: C₉H₈FNO
MW: 165.164 g/mol
Melting Pt: 125…127 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD07784308
CAS Number: 38470-28-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR